Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ascc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277057.1 | NP_001263986.1 | 28, 29 | 4749 | ||
XM_006256602.2 | XP_006256664.1 | 29, 30 | 4847 | |||
XM_006256603.2 | XP_006256665.1 | 30, 31 | 4955 | |||
XM_008772992.1 | XP_008771214.1 | 31, 32 | 5024 | |||
XM_008772993.1 | XP_008771215.1 | 29, 30 | 4868 |