Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127485.1 | NP_001120957.1 | 4 | 2375 | ||
XM_006233147.2 | XP_006233209.1 | 3 | 850 | |||
XM_006233149.2 | XP_006233211.1 | 3 | 2203 | |||
XM_008761400.1 | XP_008759622.1 | 4 | 2209 | |||
XM_008761401.1 | XP_008759623.1 | 3 | 1045 | |||
GenBank mRNA | BC099834.1 | - | 3 | 789 | ||
BC158797.1 | - | 3 | 746 |