Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abca4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107721.1 | NP_001101191.1 | 39 | 5508 | ||
XM_006233242.2 | XP_006233304.1 | 39 | 5687 |