Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Sec24d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749389.3 | XP_003749437.1 | 12, 13 | 1723 | ||
XM_006233272.2 | XP_006233334.1 | 12, 13 | 1720 | |||
GenBank mRNA | BC168946.1 | - | 5, 6 | 633 | 8366565 |