Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fpgt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199494.1 | NP_955788.1 | 4 | 990 | ||
GenBank mRNA | AJ276066.1 | - | 4 | 1025 | ||
BC168244.1 | - | 4 | 973 | 5600080 |