Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pofut1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002278.1 | NP_001002278.1 | 2 | 220 | ||
XM_006235290.2 | XP_006235352.2 | 2 | 364 | |||
GenBank mRNA | AJ781499.1 | - | 2 | 220 |