Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppfibp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107896.1 | NP_001101366.1 | 23 | 2691 | ||
XM_008763382.1 | XP_008761604.1 | 29 | 3805 | |||
XM_008763383.1 | XP_008761605.1 | 28 | 3426 | |||
XM_008763384.1 | XP_008761606.1 | 27 | 3656 | |||
XM_008763385.1 | XP_008761607.1 | 27 | 3653 | |||
XM_008763386.1 | XP_008761608.1 | 27 | 3653 | |||
XM_008763387.1 | XP_008761609.1 | 26 | 3623 | |||
XM_008763388.1 | XP_008761610.1 | 27 | 3593 | |||
XM_008763389.1 | XP_008761611.1 | 26 | 3560 | |||
XM_008763390.1 | XP_008761612.1 | 25 | 3530 | |||
XM_008763391.1 | XP_008761613.1 | 24 | 3497 | |||
XM_008763392.1 | XP_008761614.1 | 26 | 3620 | |||
XM_008763393.1 | XP_008761615.1 | 25 | 3527 |