Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC680624 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008768543.1 | XP_008766765.1 | 2 | 1458 |
Gene Symbol: Fam46b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107907.2 | NP_001101377.1 | 2 | 817 | ||
GenBank mRNA | BC158863.1 | - | 2 | 817 | 7935820 |