Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam151a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005558.1 | NP_001005558.1 | 2, 3 | 308 | ||
GenBank mRNA | BC081977.1 | - | 2, 3 | 308 | 7127557 |