Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mdm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017459.2 | NP_001017459.2 | 9 | 1292 | ||
XM_006241385.2 | XP_006241447.1 | 9 | 1329 | |||
XM_006241387.2 | XP_006241449.1 | 9 | 1316 | |||
XM_006241388.2 | XP_006241450.1 | 8 | 1299 | |||
XM_008765400.1 | XP_008763622.1 | 11 | 1606 | |||
GenBank mRNA | BC087101.1 | - | 9 | 1258 | 7192579 |