Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mon2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040176.2 | NP_001035266.2 | 34 | 5105 | ||
GenBank mRNA | FQ231480.1 | - | 3 | 367 | ||
FQ233062.1 | - | 3 | 367 |