Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Npat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108147.1 | NP_001101617.1 | 4, 5 | 521 | ||
XM_008766264.1 | XP_008764486.1 | 2, 3 | 1242 | |||
XM_008766265.1 | XP_008764487.1 | 2, 3 | 1242 | |||
XM_008766266.1 | XP_008764488.1 | 2, 3 | 1242 | |||
XM_008766267.1 | XP_008764489.1 | 2, 3 | 1242 | |||
XM_008766268.1 | XP_008764490.1 | 4, 5 | 410 |