Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Stag1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108179.1 | NP_001101649.1 | 24 | 2776 | ||
XM_006243631.2 | XP_006243693.1 | 27 | 3179 | |||
XM_006243632.2 | XP_006243694.1 | 28 | 3196 | |||
XM_008766528.1 | XP_008764750.1 | 28 | 3155 |