Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006244005.1 | XP_006244067.1 | 2 | 299 | ||
XM_006244006.2 | XP_006244068.1 | 2 | 955 | |||
XM_006244007.1 | XP_006244069.1 | 1 | 47 | |||
XM_236647.7 | XP_236647.6 | 2 | 299 |