Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inpp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012131.1 | NP_001012131.1 | 3 | 562 | ||
XM_006244899.2 | XP_006244961.1 | 4 | 782 | |||
XM_008767091.1 | XP_008765313.1 | 4 | 1523 | |||
XM_008767092.1 | XP_008765314.1 | 4 | 634 | |||
XM_008767093.1 | XP_008765315.1 | 2 | 2674 | |||
GenBank mRNA | BC086600.1 | - | 3 | 562 | 7314378 |