Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trip12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031659.1 | NP_001026829.1 | 36 | 5187 | ||
XM_006245377.2 | XP_006245439.1 | 36 | 5443 | |||
XM_006245379.2 | XP_006245441.1 | 36 | 5440 | |||
XM_006245380.2 | XP_006245442.1 | 36 | 5428 | |||
XM_006245386.1 | XP_006245448.1 | 35 | 5359 | |||
XM_008767252.1 | XP_008765474.1 | 36 | 5452 | |||
XM_008767253.1 | XP_008765475.1 | 36 | 5503 | |||
XM_008767254.1 | XP_008765476.1 | 36 | 5449 | |||
XM_008767255.1 | XP_008765477.1 | 36 | 5437 | |||
XM_008767256.1 | XP_008765478.1 | 37 | 5365 | |||
XM_008767257.1 | XP_008765479.1 | 35 | 5371 | |||
GenBank mRNA | BC088304.1 | - | 5 | 498 | ||
BC100624.1 | - | 36 | 5187 | 7930421 | ||
BC105889.1 | - | 6 | 773 |