Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcdh19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001169129.1 | NP_001162600.1 | 1 | 522 | ||
XM_006257222.2 | XP_006257284.1 | 1 | 722 | |||
XM_006257223.2 | XP_006257285.1 | 1 | 722 | |||
XM_006257224.2 | XP_006257286.1 | 1 | 722 | |||
XM_008773420.1 | XP_008771642.1 | 1 | 722 |