Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Foxp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108250.1 | NP_001101720.1 | 10 | 1139 | ||
XM_006256731.2 | XP_006256793.1 | 9 | 1202 | |||
GenBank mRNA | AB232988.1 | - | 10 | 1163 | ||
AB232989.1 | - | 9 | 1081 | |||
AB232990.1 | - | 10 | 1163 |