Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sash3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134992.1 | NP_001128464.1 | 3 | 417 | ||
XM_006257530.2 | XP_006257592.1 | 3 | 471 | |||
GenBank mRNA | BC169091.1 | - | 3 | 417 | 7933144 |