Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp11c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006257596.2 | XP_006257658.1 | 15, 16 | 1613 | ||
XM_006257597.2 | XP_006257659.1 | 15, 16 | 2626 | |||
XM_006257599.2 | XP_006257661.1 | 15, 16 | 2626 |