Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175603.2 | NP_783193.2 | 2 | 108 | ||
GenBank mRNA | AY184814.1 | - | 1 | 22 | ||
BC093598.1 | - | 2 | 108 | 7315896 |