Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akap10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114606.1 | NP_001108078.1 | 3 | 458 | ||
XM_006246509.2 | XP_006246571.1 | 3 | 2055 | |||
GenBank mRNA | BC081953.1 | - | 3 | 431 | 7122794 |