Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD1309676 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014140.1 | NP_001014162.1 | 4 | 447 | ||
XM_006252791.2 | XP_006252853.1 | 4 | 1249 | |||
GenBank mRNA | BC079275.1 | - | 4 | 447 | 7134042 |