Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata6l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014165.1 | NP_001014187.1 | 5 | 492 | ||
XM_006231237.2 | XP_006231299.1 | 5 | 451 | |||
XM_006231239.2 | XP_006231301.1 | 5 | 368 | |||
XM_006231240.2 | XP_006231302.1 | 4 | 338 | |||
XM_008760342.1 | XP_008758564.1 | 6 | 521 | |||
XM_008760343.1 | XP_008758565.1 | 6 | 522 | |||
GenBank mRNA | BC079024.1 | - | 5 | 492 | 7113918 |