Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sufu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024899.1 | NP_001020070.1 | 2 | 379 | ||
XM_006231546.2 | XP_006231608.1 | 2 | 433 | |||
GenBank mRNA | BC097447.1 | - | 2 | 379 | 7456556 |