Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: Ccar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108535.1 | NP_001102005.1 | 15 | 2075 | ||
XM_006256462.1 | XP_006256524.1 | 15 | 2021 | |||
XM_006256463.1 | XP_006256525.1 | 15 | 2025 | |||
XM_006256464.2 | XP_006256526.1 | 15 | 2022 | |||
XM_008772893.1 | XP_008771115.1 | 13 | 3040 | |||
XM_008772894.1 | XP_008771116.1 | 15 | 2019 | |||
XM_008772895.1 | XP_008771117.1 | 13 | 3039 | |||
XM_008772896.1 | XP_008771118.1 | 13 | 3040 | |||
XM_008772897.1 | XP_008771119.1 | 11 | 1460 | |||
GenBank mRNA | FQ227619.1 | - | 5 | 741 | ||
FQ233652.1 | - | 5 | 739 |
Gene Symbol: LOC100911233 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751966.3 | XP_003752014.1 | 16 | 2024 |