Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108576.1 | NP_001102046.1 | 5 | 528 | ||
XM_006233832.2 | XP_006233894.1 | 4 | 520 |