Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Api5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127379.1 | NP_001120851.1 | 3, 4 | 396 | ||
XM_006234586.1 | XP_006234648.1 | 3, 4 | 489 | |||
GenBank mRNA | BC162003.1 | - | 3, 4 | 396 | 9089120 |