Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fkbp15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763840.1 | XP_008762062.1 | 6 | 613 | ||
GenBank mRNA | FQ230831.1 | - | 5 | 540 |