Search Thermo Fisher Scientific
Gene Symbol: Zbtb48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013216.2 | NP_001013234.2 | 4 | 1074 | ||
XM_006239469.2 | XP_006239531.1 | 4 | 1083 | |||
XM_008764367.1 | XP_008762589.1 | 4 | 1112 | |||
GenBank mRNA | BC091214.1 | - | 4 | 1075 | 7383542 | |
FQ211844.1 | - | 4 | 1101 |