Gene Symbol: Stt3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170539.1 | NP_001164010.1 | 13, 14 | 2133 | ||
GenBank mRNA | AY310157.1 | - | 1589 | |||
BC166740.1 | - | 13, 14 | 2133 | 7937421 | ||
FQ210494.1 | - | 13, 14 | 2134 |