Gene Symbol: Stt3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170539.1 | NP_001164010.1 | 2, 3 | 489 | ||
GenBank mRNA | AY310157.1 | - | 224 | |||
BC166740.1 | - | 2, 3 | 489 | 7937421 | ||
FQ210494.1 | - | 2, 3 | 490 |