Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199410.2 | NP_955442.1 | 6 | 615 | ||
GenBank mRNA | BC063180.1 | - | 6 | 619 | 6920424 |