Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109526.2 | NP_001102996.2 | 7 | 630 | ||
XM_006245736.2 | XP_006245798.1 | 6 | 936 | |||
XM_008767497.1 | XP_008765719.1 | 7 | 1745 | |||
XM_008767498.1 | XP_008765720.1 | 5 | 571 |