Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdm6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108829.1 | NP_001102299.1 | 14 | 3907 | ||
GenBank mRNA | EF532390.1 | - | 9 | 1581 | ||
EF532391.1 | - | 9 | 1568 | |||
EF532392.1 | - | 9 | 1581 | |||
EF532393.1 | - | 9 | 1568 |