Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191896.1 | NP_001178825.1 | 19 | 2889 | ||
XM_008760160.1 | XP_008758382.1 | 21 | 5084 |