Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Egflam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108938.2 | NP_001102408.2 | 7, 8 | 1040 | ||
XM_006232013.2 | XP_006232075.1 | 7, 8 | 1646 | |||
XM_008760781.1 | XP_008759003.1 | 7, 8 | 4548 | |||
GenBank mRNA | BC168173.1 | - | 7, 8 | 1040 | 9030270 |