Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alg9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109000.1 | NP_001102470.1 | 8, 9 | 1021 | ||
XM_008766322.1 | XP_008764544.1 | 8, 9 | 1599 |