Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr829 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000901.1 | NP_001000901.1 | 1 | 54 | ||
XM_008763250.1 | XP_008761472.1 | 179 |