Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ece2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002815.2 | NP_001002815.2 | 6 | 647 | ||
XM_006248573.2 | XP_006248635.1 | 7 | 1146 | |||
XM_008768824.1 | XP_008767046.1 | 6 | 890 | |||
XM_008768825.1 | XP_008767047.1 | 6 | 634 |