Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: L1cam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017345.1 | NP_059041.1 | 12 | 1572 | ||
XM_008773635.1 | XP_008771857.1 | 12 | 1581 | |||
XM_008773636.1 | XP_008771858.1 | 11 | 1566 | |||
XM_008773637.1 | XP_008771859.1 | 11 | 1566 | |||
GenBank mRNA | X59149.1 | - | 12 | 1572 |