Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cacnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017346.1 | NP_059042.1 | 4 | 447 | ||
XM_006247409.2 | XP_006247471.1 | 4 | 580 | |||
XM_006247410.2 | XP_006247472.1 | 3 | 1522 | |||
XM_006247411.2 | XP_006247473.1 | 3 | 1522 | |||
GenBank mRNA | X61394.1 | - | 4 | 447 |