Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2f4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019303.1 | NP_062176.1 | 4 | 576 | ||
XM_006228585.2 | XP_006228647.1 | 5 | 684 | |||
GenBank mRNA | AF017393.1 | - | 4 | 576 | ||
BC070939.1 | - | 5 | 620 | 7099393 |