Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pfkfb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019333.1 | NP_062206.1 | 9 | 739 | ||
XM_006243840.2 | XP_006243902.1 | 8 | 698 | |||
XM_006243841.2 | XP_006243903.1 | 8 | 692 | |||
XM_006243842.2 | XP_006243904.1 | 8 | 660 | |||
XM_008766563.1 | XP_008764785.1 | 8 | 1739 | |||
GenBank mRNA | EU404101.1 | - | 8 | 708 | ||
EU404102.1 | - | 8 | 708 | |||
EU404103.1 | - | 8 | 708 | |||
M64797.1 | - | 9 | 739 |
Gene Symbol: LOC100911725 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006243892.2 | XP_006243954.1 | 670 |