Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lxn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031655.1 | NP_113843.1 | 4 | 662 | ||
GenBank mRNA | BC081763.1 | - | 4 | 648 | 7125782 | |
FQ220594.1 | - | 4 | 677 | |||
FQ229501.1 | - | 4 | 654 | |||
U40260.1 | - | 4 | 629 | |||
X76985.1 | - | 4 | 662 |