Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stx8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031656.2 | NP_113844.1 | 1, 2 | 45 | ||
XM_008767863.1 | XP_008766085.1 | 2, 3 | 269 | |||
XM_008767864.1 | XP_008766086.1 | 2, 3 | 269 | |||
XM_008767865.1 | XP_008766087.1 | 2, 3 | 269 | |||
GenBank mRNA | AF033109.1 | - | 1, 2 | 50 |