Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031658.1 | NP_113846.1 | 6 | 531 | ||
XM_006246021.2 | XP_006246083.1 | 7 | 482 | |||
XM_006246022.2 | XP_006246084.1 | 7 | 489 | |||
GenBank mRNA | BC070934.1 | - | 6 | 492 | 7098161 | |
D87351.1 | - | 6 | 531 |