Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prom1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110137.1 | NP_001103607.1 | 6 | 846 | ||
NM_021751.2 | NP_068519.2 | 7 | 913 | |||
XM_008770211.1 | XP_008768433.1 | 8 | 981 | |||
XM_008770212.1 | XP_008768434.1 | 8 | 1277 | |||
XM_008770213.1 | XP_008768435.1 | 8 | 1173 | |||
XM_008770214.1 | XP_008768436.1 | 8 | 981 | |||
XM_008770215.1 | XP_008768437.1 | 7 | 1249 | |||
XM_008770216.1 | XP_008768438.1 | 8 | 981 | |||
XM_008770217.1 | XP_008768439.1 | 8 | 981 | |||
XM_008770218.1 | XP_008768440.1 | 7 | 954 | |||
GenBank mRNA | AF263368.1 | - | 6 | 880 | ||
AF386758.1 | - | 6 | 792 | |||
AY262731.1 | - | 6 | 792 | |||
DQ871262.1 | - | 7 | 725 | |||
EV765242.1 | - | 2 | 108 |