Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdkl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021772.2 | NP_068540.1 | 5 | 732 | ||
XM_006246323.2 | XP_006246385.1 | 5 | 923 | |||
XM_006246325.2 | XP_006246387.1 | 5 | 829 | |||
XM_006246326.2 | XP_006246388.1 | 5 | 832 | |||
XM_006246327.2 | XP_006246389.1 | 5 | 908 | |||
XM_006246329.2 | XP_006246391.1 | 5 | 925 | |||
XM_006246332.2 | XP_006246394.1 | 5 | 921 | |||
XM_006246333.2 | XP_006246395.1 | 5 | 920 | |||
XM_008767705.1 | XP_008765927.1 | 5 | 922 | |||
XM_008767706.1 | XP_008765928.1 | 5 | 921 | |||
XM_008767707.1 | XP_008765929.1 | 5 | 920 | |||
GenBank mRNA | AF112183.1 | - | 5 | 732 | ||
AF112184.1 | - | 5 | 668 |