Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022260.3 | NP_071596.1 | 5 | 611 | ||
XM_008760546.1 | XP_008758768.1 | 6 | 1204 | |||
XM_008760547.1 | XP_008758769.1 | 5 | 1586 | |||
XM_008760548.1 | XP_008758770.1 | 5 | 913 | |||
XM_008760549.1 | XP_008758771.1 | 6 | 793 | |||
XM_008760550.1 | XP_008758772.1 | 5 | 721 | |||
GenBank mRNA | AF072124.1 | - | 5 | 612 | ||
BC070936.1 | - | 5 | 612 | 7099111 |